Anti-CD3 epsilon Mouse Monoclonal Antibody [clone: B-B12]
Katalog # ANTIA250502-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:CD3 epsilon
- Antigensymbol:CD3 epsilon
- Klonalitet:Monoclonal
- Klon:B-B12
- Konjugation:Unconjugated
- Flödescytometri:Yes
- Värd:Mouse
- ImmunoFluorescens:Yes
- Isotyp:IgG1 kappa
- Reaktivitet:Human
- Form:Liquid
- Gen-ID:UniprotID# P07766
- Antigen synonymer:CD3E antigen, epsilon subunit|CD3e molecule epsilon|CD3e|CD3e molecule, epsilon (CD3 TCR complex)|CD3e antigen epsilon polypeptide (TiT3 complex)|CD3e molecule, epsilon (CD3-TCR complex)|CD3E antigen epsilon polypeptide|CD3E_HUMAN|IMD18
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molekylvikt:20 kDa
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Koncentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human T cell leukemia cells
- Rening:Protein A chromatography
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Specifications
About this item
Mouse monoclonal [B-B12] antibody to CD3 epsilon for Flow Cytometry and IF with samples derived from Human.
- Validated applications: Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: CD3 epsilon
Clonality: Monoclonal
Clone: B-B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human