Anti-PGP9.5 Mouse Monoclonal Antibody [clone: UCHL1/775]
Catalog # ANTIA250273-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Epididymis luminal protein 117
- Antigen symbol:PGP9.5
- Clonality:Monoclonal
- Clone:UCHL1/775
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P09936
- Antigen synonyms:HEL 117|Neuron cytoplasmic protein 9.5|HEL S 53|Epididymis secretory protein Li 53|OTTHUMP00000218141|OTTHUMP00000218139|OTTHUMP00000218140|NDGOA|OTTHUMP00000218137
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:20 - 30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human UCHL1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [UCHL1/775] antibody to PGP9.5 for WB and IHC-P with samples derived from Human and Rat.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: PGP9.5
Clonality: Monoclonal
Clone: UCHL1/775
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Rat