Anti-TLE1 Mouse Monoclonal Antibody [clone: TLE1/2085]
Catalog # ANTIA250147-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:E(Sp1) homolog
- Antigen symbol:TLE1
- Clonality:Monoclonal
- Clone:TLE1/2085
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q04724
- Antigen synonyms:GRG1|ESG1|ESG|Enhancer of split groucho-like protein 1|TLE1_HUMAN|Enhancer of split groucho 1|TLE 1|Enhancer of split groucho like protein 1|TLE1
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:83 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 175 - 338, of human TLE1 protein. The exact sequence is proprietary
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [TLE1/2085] antibody to TLE1 for WB and IHC-P with samples derived from Human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: TLE1
Clonality: Monoclonal
Clone: TLE1/2085
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human