Anti-alpha Smooth Muscle Actin Mouse Monoclonal Antibody [clone: 1A4]
ANTIA253074-100
New Product
- Antibody type:Primary
- Antigen name:a actin
- Antigen symbol:alpha Smooth Muscle Actin
- Clonality:Monoclonal
- Clone:1A4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Rat,Mouse,Cat,Baboon,Pig,Rabbit,Monkey,Sheep,Goat,Chicken,Human,Bovine,Guinea pig,Dog
- Form:Liquid
- Gene ID:UniprotID# P62736
- Antigen synonyms:AAT6|Actin alpha 2 smooth muscle aorta|ACTA2|Actin aortic smooth muscle|ACTVS|ACTA_HUMAN|Actin, aortic smooth muscle|Alpha 2 actin|ACTSA
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:42 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:N terminal decapeptide of alpha smooth muscle isoform of Actin, conjugated to KLH
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [1A4] antibody to alpha Smooth Muscle Actin for Flow Cytometry, IF and IHC-P with samples derived from Human, Baboon, Monkey, Bovine, Porcine, Sheep, Goat, Feline, Canine, Rabbit, Mouse, Rat, Guinea Pig and Chicken.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 4 µg/ml, IHC-P: 0,25 to 0,5 µg/ml
Type: Primary
Antigen: alpha Smooth Muscle Actin
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Baboon, Monkey, Bovine, Porcine, Sheep, Goat, Cat, Dog, Rabbit, Mouse, Rat, Guinea Pig, Chicken