Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Kappa Light Chain Mouse Monoclonal Antibody [clone: KLC1278]
  ANTIA252164-100
New Product
undefined
Anti-Kappa Light Chain Mouse Monoclonal Antibody [clone: KLC1278]
  ANTIA252164-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    HCAK 1
  • Antigen symbol:
    Kappa Light Chain
  • Clonality:
    Monoclonal
  • Clone:
    KLC1278
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01601, P01834
  • Antigen synonyms:
    Immunoglobulin kappa light chain|Immunoglobulin kappa constant region|IGKC|Immunoglobulin kappa light chain constant region|Immunoglobulin InV|IGKCD|Ig kappa chain C region|HCAK1|Immunoglobulin kappa constant
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    ~22,5 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human Ig kappa chain
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [KLC1278] antibody to Kappa Light Chain for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Kappa Light Chain
Clonality: Monoclonal
Clone: KLC1278
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human