Anti-CD74 Mouse Monoclonal Antibody [clone: LN-2]
Catalog # ANTIA250771-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 74
- Antigen symbol:CD74
- Clonality:Monoclonal
- Clone:LN-2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Baboon,Human,Mouse
- Form:Liquid
- Gene ID:UniprotID# P044233
- Antigen synonyms:HG2A_HUMAN|Gamma chain of class II antigens|CD74 antigen|CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated)|CLIP|CD74 molecule|CD74 molecule, major histocompatibility complex, class II invariant chain|CD74|DHLAG
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:33 - 41 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:SU-DHL-4 lymphoma cells
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [LN-2] antibody to CD74 for Flow Cytometry and IHC-P with samples derived from Human, Baboon and Mouse.
- Validated applications: Flow cytometry, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 4 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: LN-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Baboon, Mouse