Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Amphiphysin Rabbit Polyclonal Antibody
  ANTIA11829-100
New Product
undefined
Anti-Amphiphysin Rabbit Polyclonal Antibody
  ANTIA11829-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Amp
  • Antigen symbol:
    Amphiphysin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P49418
  • Antigen synonyms:
    H1|CG8604|hiphysin (Stiff-Man syndrome with breast cancer 128kDa autoantigen)|H_HUMAN|hiphysin|hiphysin I|hiphysin (Stiff-Man syndrome with breast cancer 128kD autoantigen)|H|H 1
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    125 kDa
  • Sequence:
    MADIKTGIFAKNVQKRLNRAQEKVLQKLGKADETKDEQFEEYVQNFKRQEAEGTRLQRELRGYLAAIKGMQEASMKLTESLHEVYEPDWYGREDVKMVGEKCDVLWEDFHQKLVDGSLLTLDTYLGQFPDIKNRIAKRSRKLVDYDSARHHLEALQSSKRKDESRISKAEEEFQKAQKVFEEFNVDLQEELPSLWSRRVGFYVNTFKNVSSLEAKFHKEIAVLCHKLYEVMTKLGDQHADKAFTIQGAPS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 250 of human AMPH (NP_001626.1)
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to Amphiphysin for WB and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Amphiphysin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat