Anti-MAX (phospho Ser11) Rabbit Polyclonal Antibody
Plastic vial
Catalog # ANTIA10880-100
Supplier: Antibodies.com
Anti-MAX (phospho Ser11) Rabbit Polyclonal Antibody
Catalog # ANTIA10880-100
Supplier: Antibodies.com
Jump to:
Product Details & Documents
Rabbit polyclonal antibody to MAX (phospho Ser11) for WB with samples derived from Human.
- Recommended dilutions: WB: 1:500 to 1:2000
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: MAX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Seq:1286
- Pack type:Plastic vial
- Western blot:Yes
- Clonality:Polyclonal
- Modification:phospho Ser11
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Host:Rabbit
- Sequence:VESDE
- Concentration:Lot specific
- Molecular weight:21 kDa
- Shipping temperature:Shipped on blue ice at 4 °C
- Antigen name:MAX protein
- Cat. No.:ANTIA10880-100
- Reactivity:Human
- Gene ID:UniprotID# P61244
- Antigen synonyms:BHLHD4|Class D basic helix-loop-helix protein 4|Myc-associated factor X|Protein max
- Immunogen:A synthetic phosphorylated peptide around S11 of human MAX (NP_002373.3)
- Conjugation:Unconjugated
- Antigen symbol:MAX
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,02% sodium azide
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



