Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Guanylate kinase Rabbit Polyclonal Antibody
  ANTIA10484-100
New Product
undefined
Anti-Guanylate kinase Rabbit Polyclonal Antibody
  ANTIA10484-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ATP:GMP phosphotransferase
  • Antigen symbol:
    Guanylate kinase
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q16774
  • Antigen synonyms:
    GMP kinase|GUK1|Guanylate kinase|FLJ42686|GMK|FLJ43710|KGUA_HUMAN
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    21 kDa
  • Sequence:
    MSGPRPVVLSGPSGAGKSTLLKRLLQEHSGIFGFSVSHTTRNPRPGEENGKDYYFVTREVMQRDIAAGDFIEHAEFSGNLYGTSKVAVQAVQAMNRICVLDVDLQGVRNIKATDLRPIYISVQPPSLHVLEQRLRQRNTETEESLVKRLAAAQADMESSKEPGLFDVVIINDSLDQAYAELKEALSEEIKKAQRTGA
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 197 of human GUK1 (NP_000849.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to Guanylate kinase for WB with samples derived from human.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:200 to 1:1000

Type: Primary
Antigen: Guanylate kinase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human