Anti-CD263 Mouse Monoclonal Antibody [clone: TRAIL-R3-02] (APC (Allophycocyanin))
Catalog # ANTIA121840-100
Leverantör: Antibodies.com
New Product
Specifikationer
- Antibody type:Primary
- Antigen name:Antagonist decoy receptor for TRAIL/Apo 2L
- Antigen symbol:CD263
- Clonality:Monoclonal
- Clone:TRAIL-R3-02
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# O14798
- Antigen synonyms:Decoy Receptor 1|Lymphocyte inhibitor of TRAIL|Decoy TRAIL receptor without death domain|DcR1|MGC149501|CD263|LIT|Cytotoxic TRAIL receptor 3|DCR1 TNFR
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:0,1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein of human IgG heavy chain and extracellular domain of TRAIL - R3 (amino acids 1 - 280)
- Purification:This antibody is conjugated with APC under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifikationer
Om denna produkt
Mouse monoclonal [TRAIL-R3-02] antibody to CD263 (APC) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 5 µg/ml
Type: Primary
Antigen: CD263
Clonality: Monoclonal
Clone: TRAIL-R3-02
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human