Anti-CD253 Mouse Monoclonal Antibody [clone: 2E5] (APC (Allophycocyanin))
Catalog # ANTIA121824-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Apo 2 ligand
- Antigen symbol:CD253
- Clonality:Monoclonal
- Clone:2E5
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P50591
- Antigen synonyms:Chemokine tumor necrosis factor ligand superfamily member 10|APO 2L|CD253 antigen|Apo-2 ligand|Apo-2L|Protein TRAIL|CD253|TL2|APO2L
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:0,1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant soluble fragment, corresponding to amino acids 95 - 281, of human TRAIL
- Purification:This antibody is conjugated with APC under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [2E5] antibody to CD253 (APC) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 5 µg/ml
Type: Primary
Antigen: CD253
Clonality: Monoclonal
Clone: 2E5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human