Anti-Ubiquitin Goat Polyclonal Antibody
ANTIA121644-600
New Product
- Antibody type:Primary
- Antigen name:Epididymis secretory protein Li 50
- Antigen symbol:Ubiquitin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Monkey,Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:UBA 80|RPS27A|UBA52|FLJ25987|MGC8385|RPS 27A|Polyubiquitin B|HEL S 50|UBA 52
- Storage buffer:Supplied in Phosphate Buffered Saline with 20% Glycerol and 0,05% Sodium Azide
- Sequence:MQIFVKTLTGKTITLEVEPSDTIENVKAKIQDKEGIPPDQQRLIFAGKQLEDGRTLSDYNIQKESTLHLVLRLRGG
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
- Concentration:3 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant peptide corresponding to human Ubiquitin, expressed in and purified from E. coli
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:600 µG
Goat polyclonal antibody to Ubiquitin for WB with samples derived from human, rat, mouse, canine and monkey.
- Validated applications: WB
- Recommended dilutions: WB: 1:250 to 1:2000
Type: Primary
Antigen: Ubiquitin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Rat;Mouse;Dog;Monkey