Anti-CD15 Mouse Monoclonal Antibody [clone: MMA] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA122115-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:3-FAL
- Antigen symbol:CD15
- Clonality:Monoclonal
- Clone:MMA
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:CD15|FUC-TIV|Alpha-3-fucosyltransferase|ELAM 1 ligand|ELFT|fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific)|ELAM-1 ligand fucosyltransferase|FCT3A|ELAM ligand fucosyltransferase
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:U937 histiocytic lymphoma cells
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MMA] antibody to CD15 (FITC) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: MMA
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human