Anti-CD172b Mouse Monoclonal Antibody [clone: B4B6] (PE (Phycoerythrin))
Catalog # ANTIA121865-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD172 antigen like family member B
- Antigen symbol:CD172b
- Clonality:Monoclonal
- Clone:B4B6
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# O00241
- Antigen synonyms:SIRP BETA 1|CD172b antigen|1/CD172b|Signal regulatory protein beta 1|SIRPß|DKFZp686A05192|SIRP beta 1 isoform 3|CD172b
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:NIH-3T3 cells transfected with human CD172b
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [B4B6] antibody to CD172b (PE) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD172b
Clonality: Monoclonal
Clone: B4B6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human