Order Entry
Sweden
ContactUsLinkComponent
Anti-I-FABP Mouse Monoclonal Antibody [clone: FABP2/7670]
Anti-I-FABP Mouse Monoclonal Antibody [clone: FABP2/7670]
Catalog # ANTIA316517-100
Supplier:  ANTIBODIES.COM
New Product
Anti-I-FABP Mouse Monoclonal Antibody [clone: FABP2/7670]
Catalog # ANTIA316517-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FABP 2
  • Antigen symbol:
    I-FABP
  • Clonality:
    Monoclonal
  • Clone:
    FABP2/7670
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P12104
  • Antigen synonyms:
    Fatty acid binding protein 2 intestinal|Fatty acid binding protein|Fatty acid-binding protein 2|Fatty acid binding protein intestinal|Fatty acid-binding protein|I FABP|FABPI_HUMAN|FABP2|FABPI
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    15,21 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant full length human FABP2 protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [FABP2/7670] antibody to I-FABP for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: I-FABP
Clonality: Monoclonal
Clone: FABP2/7670
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human