Anti-HDAC7 Mouse Monoclonal Antibody [clone: PCRP-HDAC7-1B6]
ANTIA316454-100
New Product
- Antibody type:Primary
- Antigen name:DKFZP586J0917
- Antigen symbol:HDAC7
- Clonality:Monoclonal
- Clone:PCRP-HDAC7-1B6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q8WUI4
- Antigen synonyms:FLJ99588|HDAC7|HDAC7_HUMAN|HD7a|HDAC 7|HDAC 7A|HD7|HDAC7A|HD 7a
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:105 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant full-length human HDAC7 protein.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PCRP-HDAC7-1B6] antibody to HDAC7 for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: HDAC7
Clonality: Monoclonal
Clone: PCRP-HDAC7-1B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human