Anti-Glypican 3 Rabbit Recombinant Antibody [clone: GPC3/8363R]
ANTIA316424-100
New Product
- Antibody type:Primary
- Antigen name:DGSX
- Antigen symbol:Glypican 3
- Clonality:Recombinant
- Clone:GPC3/8363R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P51654
- Antigen synonyms:Gpc3|Heparan sulphate proteoglycan|Glypican 3|Glypican-3 [Precursor]|Intestinal protein OCI 5|GTR2-2|GTR2 2|GPC3_HUMAN|Glypican proteoglycan 3
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 400-550) of human GPC3 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [GPC3/8363R] antibody to Glypican 3 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Glypican 3
Clonality: Recombinant
Clone: GPC3/8363R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human