Order Entry
Sweden
ContactUsLinkComponent
Anti-GATA4 Mouse Monoclonal Antibody [clone: PCRP-GATA4-1A7]
Anti-GATA4 Mouse Monoclonal Antibody [clone: PCRP-GATA4-1A7]
  ANTIA316379-100
New Product
Anti-GATA4 Mouse Monoclonal Antibody [clone: PCRP-GATA4-1A7]
  ANTIA316379-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    ASD2
  • Antigen symbol:
    GATA4
  • Clonality:
    Monoclonal
  • Clone:
    PCRP-GATA4-1A7
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P43694
  • Antigen synonyms:
    Transcription factor GATA-4|Transcription factor GATA4|GATA 4|GATA binding protein 4|GATA4|GATA-binding factor 4|MGC126629|Transcription factor GATA 4|GATA4_HUMAN
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    44,57 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 262-321) of human GATA4 protein.
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [PCRP-GATA4-1A7] antibody to GATA4 for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells

Type: Primary
Antigen: GATA4
Clonality: Monoclonal
Clone: PCRP-GATA4-1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human