Anti-CD99 Mouse Monoclonal Antibody [clone: MIC2/7866]
Katalog # ANTIA316149-100
Leverantör: ANTIBODIES.COM
New Product
Specifikationer
- Antibody type:Primary
- Antigen name:CD99
- Antigen symbol:CD99
- Clonality:Monoclonal
- Clone:MIC2/7866
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P14209
- Antigen synonyms:CD99|CD99_HUMAN|Antigen identified by monoclonal antibodies 12E7, F21 and O13|12E7|CD99 molecule|Cell surface antigen O13|Antigen identified by monoclonal 12E7, Y homolog|Cell surface antigen 12E7|CD99 antigen|Cell surface antigen HBA 71
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:27 - 32 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-185) of human MIC2 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifikationer
Om denna produkt
Mouse monoclonal [MIC2/7866] antibody to CD99 for WB and IHC-P with samples derived from human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: MIC2/7866
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human