Anti-CD63 Mouse Monoclonal Antibody [clone: LAMP3/7369]
ANTIA316109-100
New Product
- Antibody type:Primary
- Antigen name:CD 63
- Antigen symbol:CD63
- Clonality:Monoclonal
- Clone:LAMP3/7369
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08962
- Antigen synonyms:CD63_HUMAN|gp55|CD63|CD63 molecule|Granulophysin|CD63 antigen (melanoma 1 antigen)|CD63 antigen|LAMP 3|CD63 antigen melanoma 1 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:26 kDa (core protein)/30 - 60 kDa (glycosylated)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 100-200) of human CD63 protein. The exact sequence is proprietary.
- Tested applications:Functional Studies, IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [LAMP3/7369] antibody to CD63 for FUNC, IP, flow cytometry, IF and IHC-P with samples derived from human.
- Validated applications: FUNC, IP, Flow cytometry, IF, IHC-P
- Recommended dilutions: Functional studies, IP: 1 to 2 µg per 100 to 500 µg of total protein (1 ml of cell lysate), Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: LAMP3/7369
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human