Anti-Uroplakin Ib/UPIb Rabbit Recombinant Antibody [clone: UPK1B/8976R]
ANTIA315772-100
New Product
- Antibody type:Primary
- Antigen symbol:Uroplakin Ib/UPIb
- Clonality:Recombinant
- Clone:UPK1B/8976R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# O75841
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 109-229) of human Uroplakin 1B (UPK1B) protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [UPK1B/8976R] antibody to Uroplakin Ib/UPIb for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Uroplakin Ib/UPIb
Clonality: Recombinant
Clone: UPK1B/8976R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human