Order Entry
Sweden
ContactUsLinkComponent
Anti-Smoothelin Rabbit Recombinant Antibody [clone: SMTN/2320R]
Anti-Smoothelin Rabbit Recombinant Antibody [clone: SMTN/2320R]
Catalog # ANTIA315661-100
Supplier:  ANTIBODIES.COM
New Product
Anti-Smoothelin Rabbit Recombinant Antibody [clone: SMTN/2320R]
Catalog # ANTIA315661-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FLJ35168
  • Antigen symbol:
    Smoothelin
  • Clonality:
    Recombinant
  • Clone:
    SMTN/2320R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P53814
  • Antigen synonyms:
    Smoothelin|FLJ38597|FLJ35365|Smtn|SMTN_HUMAN|SMSMO|FLJ35620
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Molecular weight:
    59 kDa/110 kDa (short/long isoforms)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human Smoothelin (SMTN) protein
  • Tested applications:
    IHC
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [SMTN/2320R] antibody to Smoothelin for IHC with samples derived from human.

  • Validated applications: IHC
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Smoothelin
Clonality: Recombinant
Clone: SMTN/2320R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human