Anti-S100 beta Mouse Monoclonal Antibody [clone: S100B/4152]
ANTIA315565-100
New Product
- Antibody type:Primary
- Antigen name:NEF
- Antigen symbol:S100 beta
- Clonality:Monoclonal
- Clone:S100B/4152
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P04271
- Antigen synonyms:S-100 protein beta chain|S100 calcium binding protein beta (neural)|S 100 calcium binding protein beta chain|S 100 protein beta chain|Protein S100 B|S100|S-100 protein subunit beta|Protein S100-B|S100 beta
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:10 - 12 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-92) of human S100B protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [S100B/4152] antibody to S100 beta for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: S100 beta
Clonality: Monoclonal
Clone: S100B/4152
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human