Order Entry
Sweden
ContactUsLinkComponent
Anti-Ret Mouse Monoclonal Antibody [clone: RET/8787]
Anti-Ret Mouse Monoclonal Antibody [clone: RET/8787]
Catalog # ANTIA315545-100
Supplier:  ANTIBODIES.COM
New Product
Anti-Ret Mouse Monoclonal Antibody [clone: RET/8787]
Catalog # ANTIA315545-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    C ret
  • Antigen symbol:
    Ret
  • Clonality:
    Monoclonal
  • Clone:
    RET/8787
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2c kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P07949
  • Antigen synonyms:
    Cadherin related family member 16|CDHF 12|Cadherin family member 12|Hydroxyaryl protein kinase|ELKS Fusion gene|CDHF12|CDHR16|HSCR1|HSCR 1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Molecular weight:
    150 kDa (precursor)/170 kDa (Mature)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 702-848) of human RET protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [RET/8787] antibody to Ret for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Ret
Clonality: Monoclonal
Clone: RET/8787
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human