Anti-PSGL-1 Rabbit Recombinant Antibody [clone: PSGL1/8192R]
Catalog # ANTIA315535-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 162
- Antigen symbol:PSGL-1
- Clonality:Recombinant
- Clone:PSGL1/8192R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q14242
- Antigen synonyms:CD162 antigen|P selectin glycoprotein ligand 1|Cutaneous lymphocyte associated associated antigen|CLA|P-selectin glycoprotein ligand 1|CD162|PSGL-1|P selectin glycoprotein ligand 1 precursor|PSGL 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:43-120 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human SELPLG (CD162) protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [PSGL1/8192R] antibody to PSGL-1 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: PSGL-1
Clonality: Recombinant
Clone: PSGL1/8192R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human