Anti-ID4 Mouse Monoclonal Antibody [clone: PCRP-ID4-1A10]
Catalog # ANTIA315128-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:bHLHb27
- Antigen symbol:ID4
- Clonality:Monoclonal
- Clone:PCRP-ID4-1A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P47928
- Antigen synonyms:Class B basic helix-loop-helix protein 27|Inhibitor of DNA binding 4 dominant negative helix loop helix protein|DNA binding protein inhibitor ID 4|DNA-binding protein inhibitor ID-4|Inhibitor of DNA binding 4|DNA binding protein inhibitor ID4|ID4_HUMAN|IDB4
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:16,6 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 62-112) of human ID4 protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-ID4-1A10] antibody to ID4 for ELISA, IP and WB with samples derived from human.
- Validated applications: ELISA, IP, WB
- Recommended dilutions: IP: 1 to 2 µg per 100 to 500 µg of total protein (1 ml of cell lysate), WB: 1 to 2 µg/ml
Type: Primary
Antigen: ID4
Clonality: Monoclonal
Clone: PCRP-ID4-1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human