Anti-M-CSF Recombinant Recombinant Antibody [clone: DMC437]
Supplier: Antibodies.com
New Product
Certificates
About this item
Recombinant chimeric monoclonal [DMC437] antibody to M-CSF for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: M-CSF
Clonality: Recombinant
Clone: DMC437
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ANTIA318741-500
- ANTIA318741-100
- Antigen symbol:
- M-CSF
- M-CSF
- Antigen name:
- Colony stimulating factor 1
- Colony stimulating factor 1
- Antigen synonyms:
- Colony stimulating factor 1 (macrophage)|CSF1_HUMAN|CSF-1|M CSF|Lanimostim|CSF 1|Csfm|CSF1|Colony stimulating factor macrophage specific
- Colony stimulating factor 1 (macrophage)|CSF1_HUMAN|CSF-1|M CSF|Lanimostim|CSF 1|Csfm|CSF1|Colony stimulating factor macrophage specific
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- DMC437
- DMC437
- Reactivity:
- Human
- Human
- Host:
- Recombinant
- Recombinant
- Gene ID:
- UniprotID# P09603
- UniprotID# P09603
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilised
- Lyophilised
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% trehalose is added as a protectant before lyophilisation.
- Storage temperature:
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together