Anti-Prolactin Receptor Mouse Monoclonal Antibody [clone: PRLR/742]
Katalog # ANTIA249741-100
Leverantör: ANTIBODIES.COM
New Product
Specifikationer
- Antibody type:Primary
- Antigen name:AI987712
- Antigen symbol:Prolactin Receptor
- Clonality:Monoclonal
- Clone:PRLR/742
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P16471
- Antigen synonyms:Delta 4-delta 7/11 truncated prolactin receptor|hPRLrI|Lactogen receptor|MFAB|CPRLP|hPRL receptor|Delta 4-SF1b truncated prolactin receptor|HPRL|CLONE SPM213
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:70 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human Prolactin receptor
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifikationer
Om denna produkt
Mouse monoclonal [PRLR/742] antibody to Prolactin Receptor for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Prolactin Receptor
Clonality: Monoclonal
Clone: PRLR/742
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human