Anti-MUC4 Mouse Monoclonal Antibody [clone: MUC4/3105]
ANTIA249429-100
New Product
- Antibody type:Primary
- Antigen name:Ascites sialoglycoprotein
- Antigen symbol:MUC4
- Clonality:Monoclonal
- Clone:MUC4/3105
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q99102
- Antigen synonyms:Ascites sialoglycoprotein 2|ASGP-1|MUC 4|Ascites sialoglycoprotein 1|HSA276359|MUC4|ASGP|MUC-4|ASGP-2
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:235 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 1730-1864, of human MUC4 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [MUC4/3105] antibody to MUC4 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: MUC4
Clonality: Monoclonal
Clone: MUC4/3105
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human