Anti-CD11b Mouse Monoclonal Antibody [clone: ITGAM/3340]
ANTIA249064-100
New Product
- Antikroppstyp:Primär
- Antikroppsnamn:antigen CD11b (p170)
- Antigensymbol:CD11b
- Klonalitet:Monoclonal
- Klon:ITGAM/3340
- Konjugation:Unconjugated
- Värd:Mouse
- ImmunKemi:Yes
- Isotyp:IgG2a kappa
- Reaktivitet:Human
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# P11215
- Antigen synonymer:CD49d|Antigen CD11b p170|Cell surface glycoprotein MAC-1 subunit alpha|CD11 antigen-like family member B|Complement component 3 receptor 3 subunit|Complement Component Receptor 3 Alpha|CD11b/CD18|CD11b|CD11 antigen like family member B
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molekylvikt:95 kDa/170 kDa
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Koncentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 941-1074, of human ITGAM protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Rening:Protein A/G chromatography
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Mouse monoclonal [ITGAM/3340] antibody to CD11b for WB and IHC-P with samples derived from human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: ITGAM/3340
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human