Anti-Annexin A1 Mouse Monoclonal Antibody [clone: CPTC-ANXA1-1]
Catalog # ANTIA248814-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Annexin 1
- Antigen symbol:Annexin A1
- Clonality:Monoclonal
- Clone:CPTC-ANXA1-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04083
- Antigen synonyms:ANX 1|Annexin I (lipocortin I)|Annexin I|ANX1|ANX A1|Annexin-1|AnnexinA1|AnnexinI|Annexin A1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:38 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human Annexin A1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [CPTC-ANXA1-1] antibody to Annexin A1 for IP, WB and IHC-P with samples derived from human.
- Validated applications: IP, WB, IHC-P
- Recommended dilutions: IP: 2 to 4 µg/mg protein, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Annexin A1
Clonality: Monoclonal
Clone: CPTC-ANXA1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human