Anti-Histone H1 Mouse Recombinant Antibody [clone: r1415-1]
Catalog # ANTIA248806-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:dJ221C16.2
- Antigen symbol:Histone H1
- Clonality:Recombinant
- Clone:r1415-1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3005
- Antigen synonyms:dJ221C16.5|H1 histone family member N testis specific|H1 histone family member 3|H1 histone family|H1 0 histone|H1 histone family member 0|DmeH1|H1 histone family member 5|H1 histone family member 1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:~30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human Histone H1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [r1415-1] antibody to Histone H1 for WB, flow cytometry, IF and IHC-P with samples derived from human, mouse and rat.
- Validated applications: WB, Flow cytometry, IF, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Histone H1
Clonality: Recombinant
Clone: r1415-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat