Anti-PD-L1 Mouse Monoclonal Antibody [clone: PDL1/2745]
ANTIA248772-100
New Product
- Antibody type:Primary
- Antigen name:B7 H
- Antigen symbol:PD-L1
- Clonality:Monoclonal
- Clone:PDL1/2745
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9NZQ7
- Antigen synonyms:B7-H1|B7 H1|B7H1|CD 274|CD274 molecule|CD274|B7 homolog 1|B7H|CD274 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:37 - 50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 39-191, of human CD274 protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PDL1/2745] antibody to PD-L1 for ELISA with samples derived from human.
- Validated applications: ELISA
Type: Primary
Antigen: PD-L1
Clonality: Monoclonal
Clone: PDL1/2745
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human