Order Entry
Sweden
ContactUsLinkComponent
 
Anti-beta Catenin Mouse Monoclonal Antibody [clone: CTNNB1/1507]
  ANTIA248298-100
New Product
undefined
Anti-beta Catenin Mouse Monoclonal Antibody [clone: CTNNB1/1507]
  ANTIA248298-100
New Product

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    beta Catenin
  • Klonalitet:
    Monoclonal
  • Klon:
    CTNNB1/1507
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG1 kappa
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Form:
    Liquid
  • Gen-ID:
    UniprotID# P35222
  • Antigen synonymer:
    CTNB1_HUMAN|CHBCAT|CATNB|Catenin beta 1|Catenin (cadherin associated protein), beta 1, 88kDa|Beta-catenin|Catenin beta-1|CTNNB|Cadherin associated protein
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molekylvikt:
    92 kDa
  • Förvaringstemperatur:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Koncentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment of human beta Catenin protein
  • Tested applications:
    IHC-P
  • Rening:
    Protein A/G chromatography
  • Förpackningstyp:
    Plastic vial
  • Förp.:
    100 µG

 

 

Mouse monoclonal [CTNNB1/1507] antibody to beta Catenin for IHC-P with samples derived from human, mouse and rat.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: beta Catenin
Clonality: Monoclonal
Clone: CTNNB1/1507
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat