Anti-LI Cadherin Mouse Monoclonal Antibody [clone: CDH17/2616]
ANTIA248073-100
New Product
- Antibody type:Primary
- Antigen name:BILL-cadherin
- Antigen symbol:LI Cadherin
- Clonality:Monoclonal
- Clone:CDH17/2616
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q12864
- Antigen synonyms:CDH16|CAD17_HUMAN|Cadherin-17|cadherin, liver-intestine|cadherin 17, LI cadherin (liver-intestine)|Cadherin 16, formerly|CDH16, formerly|Cadherin 16|Cadherin 17
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:120 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 242-418, of human Cadherin 17 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [CDH17/2616] antibody to LI Cadherin for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: LI Cadherin
Clonality: Monoclonal
Clone: CDH17/2616
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human