Anti-FNIP1 Rabbit Polyclonal Antibody
Catalog # ANTIA304955-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:FLCN-interacting protein
- Antigensymbol:FNIP1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:UniprotID# Q8TF40
- Antigen synonymer:KIAA1961|FNIP1|Folliculin interacting protein 1
- Storage buffer:Supplied in phosphate buffered saline with 50% glycerol and 0,09% sodium azide
- Molekylvikt:~130 kDa
- Förvaringstemperatur:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Koncentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide from the mid - protein of Human FNIP1.
- Tested applications:ICC/IF, IHC
- Rening:peptide. affinity purification
- Förpackningstyp:Plastic vial
- Förp.:100 µG
Specifications
About this item
Rabbit polyclonal antibody to FNIP1 for WB, ICC/IF and IHC with samples derived from human, mouse and rat.
- Validated applications: WB, ICC/IF, IHC
- Recommended dilutions: WB: 1:1000, ICC/IF: 1:100, IHC: 1:50
Type: Primary
Antigen: FNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat