Anti-alpha B Crystallin Rabbit Polyclonal Antibody
Catalog # ANTIA304739-200
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:AACRYA
- Antigen symbol:alpha B Crystallin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P02511
- Antigen synonyms:Crystallin alpha polypeptide 2|Alpha-crystallin B chain|Crystallin alpha B|CRYA2|Cryab|CRYAB_HUMAN|Alpha(B)-crystallin|Alpha B Crystallin|Alpha crystallin B chain
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~22 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to human alpha B crystallin conjugated to KLH.
- Tested applications:IHC, ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:200 µl
Specifications
About this item
Rabbit polyclonal antibody to alpha B Crystallin for WB, IHC and ICC/IF with samples derived from human, mouse, rat, bovine and chicken.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:5000, ICC/IF: 1:120
Type: Primary
Antigen: alpha B Crystallin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Bovine;Chicken