Order Entry
Sweden
ContactUsLinkComponent
 
Anti-Aly/Ref Rabbit Monoclonal Antibody [Clone: ARC0959]
  ANTIA305535-100
New Product
undefined
Anti-Aly/Ref Rabbit Monoclonal Antibody [Clone: ARC0959]
  ANTIA305535-100
New Product

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Aly / Ref
  • Clonality:
    Monoclonal
  • Clone:
    ARC0959
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q86V81
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    27 kDa
  • Sequence:
    DALKAMKQYNGVPLDGRPMNIQLVTSQIDAQRRPAQSVNRGGMTRNRGAGGFGGGGGTRRGTRGGARGRGRGAGRNSKQQLSAEELDAQLDAYNARMDTS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 158 - 257 of human THOC4/ALYREFREF (Q86V81).
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [ARC0959] antibody to Aly/Ref for WB and IHC with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: Aly/Ref
Clonality: Monoclonal
Clone: ARC0959
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat