Anti-ATG5 Rabbit Polyclonal Antibody
ANTIA304902-100
New Product
- Antibody type:Primary
- Antigen name:APG 5
- Antigen symbol:ATG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9H1Y0
- Antigen synonyms:APG5|APG5-like|APG5L|Apoptosis-specific protein|APG5 autophagy 5 like|APG5 like|APG 5L|APG5L/ATG5|Apoptosis specific protein
- Storage buffer:Supplied in phosphate buffered saline with 50% glycerol and 0,09% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide from the N - terminal of human ATG5.
- Tested applications:ICC/IF
- Purification:Peptide affinity purification
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to ATG5 for WB and ICC/IF with samples derived from human.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: ATG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human