Anti-Rat IgG Fc Goat Polyclonal Antibody
Catalog # ANTIA294712-500
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Secondary
- Antigen symbol:Rat IgG Fc
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Form:Liquid
- Antibody fragment:Whole Molecule
- Storage buffer:Supplied in phosphate buffered saline with 0,09% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:IHC, ICC
- Purification:Antigen affinity purification
- Pack type:Plastic vial
- Pk:500 µG
Specifications
About this item
Goat anti-rat IgG Fc secondary antibody for WB, IHC, ICC and ELISA with samples derived from rat.
- Validated applications: WB, IHC, ICC, ELISA
- Recommended dilutions: ELISA: 1:1000 to 1:20000; ELISA (capture antibody): 1:50 to 1:250, ICC: 1:100 to 1:1000, IHC: 1:100 to 1:1000, WB: 1:1000 to 1:20000
Type: Secondary
Antigen: Rat IgG Fc
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat