Anti-Pig IgG Fc Goat Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
ANTIA294482-500
New Product
- Antibody type:Secondary
- Antigen symbol:Pig IgG Fc
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig
- Western blot:Yes
- Form:Liquid
- Antibody fragment:Whole Molecule
- Storage buffer:Supplied in 50 mM HEPES, pH 7,1, with 0,1 M sodium chloride, 1 mM magnesium chloride, 0,1 mM zinc chloride, 0,2% BSA, and 0,09% sodium azide
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze!
- Concentration:0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:IHC, ICC
- Purification:Antigen affinity purification
- Pack type:Plastic vial
- Pk:500 µG
Goat anti-pig IgG Fc (AP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from pig.
- Validated applications: WB, IHC, ICC, ELISA
- Recommended dilutions: ELISA: 1:5000 to 1:50000, ICC: 1:100 to 1:1000, IHC: 1:100 to 1:1000, WB: 1:5000 to 1:50000
Type: Secondary
Antigen: Pig IgG Fc
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Pig