Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Myelin Expression Factor 2
- Description:Human myelin expression factor 2 ELISA kit
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Cross reactivity:Human Myelin Expression Factor 2 ELISA kit exhibits high specificity and excellent specificity for the detection of human Myelin Expression Factor 2. No significant cross-reactivity or interference between Myelin Expression Factor 2 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:31,25 - 2000 pg/ml
- Storage temperature:4 °C
- Sample volume:100 μl
- Sensitivity:18,75 pg/ml
- Regulatory status:RUO
Specifications
About this item
Human Myelin Expression Factor 2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Myelin Expression Factor 2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 31,25 to 2000 pg/ml
- Sensitivity: 18,75 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human Myelin Expression Factor 2 employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Myelin Expression Factor 2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Myelin Expression Factor 2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Myelin Expression Factor 2 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Myelin Expression Factor 2 Antibody, which binds the captured Myelin Expression Factor 2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Myelin Expression Factor 2 captured in each well. The concentration of Myelin Expression Factor 2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.