Order Entry
Sweden
ContactUsLinkComponent
 
Human N-Myc Interactor/NMI ELISA Kit
  ANTIA312888-96
New Product
 :  
undefined
Human N-Myc Interactor/NMI ELISA Kit
  ANTIA312888-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    N-Myc Interactor/NMI
  • Description:
    Human N-Myc Interactor/NMI ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    100 - 1600 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    7,44 ng/L
  • Regulatory status:
    RUO

 

 

Human N-Myc Interactor/NMI ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human N-Myc Interactor/NMI in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 100 to 1600 ng/L
  • Sensitivity: 7,44 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human N-Myc Interactor/ nmI ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human N-Myc Interactor/ nmI in serum, plasma or other biological fluids. An antibody specific for N-Myc Interactor/ nmI has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the N-Myc Interactor/ nmI present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-N-Myc Interactor/ nmI antibody, which also binds the N-Myc Interactor/ nmI present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-N-Myc Interactor/ nmI antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-N-Myc Interactor/ nmI antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of N-Myc Interactor/ nmI captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of N-Myc Interactor/ nmI in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.