Order Entry
Sweden
ContactUsLinkComponent
Rat Flt3 Ligand/Flt3L ELISA Kit
Rat Flt3 Ligand/Flt3L ELISA Kit
  ANTIA79825-96
New Product
 :  
undefined
Rat Flt3 Ligand/Flt3L ELISA Kit
  ANTIA79825-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Flt3 Ligand/Flt3L
  • Description:
    Rat Flt3 ligand/Flt3L ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Rat Flt3 Ligand/Flt3L ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Flt3 Ligand/Flt3L. No significant cross-reactivity or interference between Flt3 Ligand/Flt3L and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    1,563 - 100 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,938 ng/ml
  • Regulatory status:
    RUO

 

 

Rat Flt3 Ligand/Flt3L ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Flt3 Ligand/Flt3L in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 1,563 to 100 ng/ml
  • Sensitivity: 0,938 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Flt3 Ligand/Flt3L ELISA kit (A79825) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Flt3 Ligand/Flt3L in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Flt3 Ligand/Flt3L has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Flt3 Ligand/Flt3L present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Flt3 Ligand/Flt3L Antibody, which binds the captured Flt3 Ligand/Flt3L present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Flt3 Ligand/Flt3L captured in each well. The concentration of Flt3 Ligand/Flt3L can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.