Order Entry
Sweden
ContactUsLinkComponent
Triethanolamine (trolamine) BP, Ph. Eur., USP-NF, pharma grade
Triethanolamine (trolamine) BP, Ph. Eur., USP-NF, pharma grade
Supplier:  PanReac AppliChem
New Product
Certificates
Triethanolamine (trolamine) BP, Ph. Eur., USP-NF, pharma grade
Supplier:  PanReac AppliChem
New Product

Specification Test Results

  • Assay (Acidim.):
    99.0 - 103.0%
  • Identity according to Pharmacopoeias:
    passes test
  • Density 20/20:
    1.120 - 1.130
  • Density 25/25:
    1.120 - 1.128
  • Refractive Index n 20/D:
    1.482 - 1.485
  • Appearance of solution:
    passes test
  • Residue on ignition (as SO4):
    0.05%
  • Residual solvents (Ph.Eur/USP):
    passes test
  • Ethanolamine:
    0.1%
  • Diethanolamine:
    0.5%
  • Total impurities:
    1.0%
  • N-Nitrosodiethanolamine:
    24 ppb
  • Water (H2O):
    0.5%

Specification Test Results