Order Entry
Sweden
ContactUsLinkComponent
 
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (APC)
 
undefined
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (APC)

 

  • Catalog No:
  • STMC100-0300
  • STMC100-0299
  • STMC100-0298
  • Antigen name:
  • CD4
  • CD4
  • CD4
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • RPA-T4
  • RPA-T4
  • RPA-T4
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 920
  • 920
  • 920
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa

 

Frequently Bought Together