Order Entry
Sweden
ContactUsLinkComponent
Human Recombinant FGF-7, ACF
Human Recombinant FGF-7, ACF
Certificates
undefined
Human Recombinant FGF-7, ACF

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    FGF-7, ACF
  • Purity:
    ≥90%

Specifications

Frequently Bought Together