Order Entry
Sweden
ContactUsLinkComponent
 
Human Recombinant HGF (CHO-expressed)
 
undefined
Human Recombinant HGF (CHO-expressed)

 

  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Protein synonyms:
    DFNB39|FTCF|Fibroblast-derived tumor cytotoxic factor|Hepapoietin A|HPTA|Lung fibroblast-derived mitogen|Scatter factor|SF
  • Protein/peptide name:
    HGF
  • Purity:
    ≥95%

 

Frequently Bought Together