Human Recombinant TGF-alpha
Transforming growth factor-alpha (TGF-α) a member of the epidermal growth factor (EGF) family that is expressed in normal epithelial cells, monocytes, macrophages, brain cells, keratinocytes, and cancer cells. TGF-α binds EGF receptor (EGFR) and activates tyrosine kinase signaling. In epidermal and epithelial cells, this results in proliferation and differentiation. TGF-α is not structurally or genetically related to TGF-β, and the two ligands act through different signaling pathways. In vitro, TGF-α was shown to stimulate anchorage-independent growth (Singh and Coffey).
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
- Protein/peptide type:Recombinant
- Species:Human
- Protein/peptide name:TGF-alpha
- Purity:≥95%
Frequently Bought Together