0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Freeze Dryers
Accessories for LSCplus Controller
Accessories for LSCplus Controller
Catalog # CHRS120862
Supplier:
CHRIST MARTIN
Expand image
Accessories for LSCplus Controller
Catalog # CHRS120862
Supplier:
CHRIST MARTIN
Specifications
Description:
Fan for circulation of cold air, 200 mm, integrated into (CHRS120861) shelves
Pk:
1
For:
LSCplus controller
Specifications
About this item
Product Details
Product Details